Skip to Content
Merck
All Photos(1)

Key Documents

SML2484

Sigma-Aldrich

AZD3839

≥98% (HPLC)

Synonym(s):

(1S)-1-[2-(Difluoromethyl)-4-pyridinyl]-4-fluoro-1-[3-(5-pyrimidinyl)phenyl]-1H-isoindol-3-amine, (S)-1-[2-(Difluoromethyl)pyridin-4-yl]-4-fluoro-1-[3-(pyrimidin-5-yl)phenyl]-1H-isoindol-3-amine, AZD 3839, AZD-3839

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H16F3N5
CAS Number:
Molecular Weight:
431.41
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

NC1=N[C@@](C2=CC=CC(C3=CN=CN=C3)=C2)(C4=CC(C(F)F)=NC=C4)C5=CC=CC(F)=C51

InChI

1S/C24H16F3N5/c25-19-6-2-5-18-21(19)23(28)32-24(18,17-7-8-31-20(10-17)22(26)27)16-4-1-3-14(9-16)15-11-29-13-30-12-15/h1-13,22H,(H2,28,32)/t24-/m0/s1

InChI key

MRXBCEQZNKUUIP-DEOSSOPVSA-N

Biochem/physiol Actions

AZD3839 is a potent and selective BACE1 inhibitor (Ki = 26.1 nM/BACE1, 372.4 nM/BACE2, >25 μM/Cathepsin D by cell-free TR-FRET; hERG IC50 = 4.8 μM) that effectively suppresses cellular BACE1 activity in neuron cultures (sAPPβ release IC50 = 16.7 nM/SH-SY5Y; Aβ40 release IC50 = 4.8 nM/SH-SY5Y AAP695wt, 32.2 nM/N2A, 24.8 nM/guinea pig cortical neurons, 51 nM/murine cortical neurons) and exhibits in vivo Aβ40-, Aβ42-, sAPPβ-reducing efficacy (80 & 160 μmol/kg mouse, p.o.; 100 & 200 μmol/kg guinea pigs, p.o., 2.5 & 20 μmol/kg cynomolgus monkeys, i.v.) with good pharmacokinetic properties and oral availability.
Orally available, potent and selective BACE1 inhibitor with in vivo Aβ40-, Aβ42-, sAPPβ-reducing efficacy in mice, guinea pigs, and monkeys.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service