Skip to Content
Merck
All Photos(1)

Key Documents

SML1641

Sigma-Aldrich

WWL70

≥98% (HPLC)

Synonym(s):

4′-carbamoylbiphenyl-4-yl methyl(3-(pyridin-4-yl)benzyl)carbamate, N-methyl-N-[[3-(4-pyridinyl)phenyl]methyl]-4′-(aminocarbonyl)[1,1′-biphenyl]-4-yl ester carbamic acid, WWL-70

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H23N3O3
CAS Number:
Molecular Weight:
437.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CN(C(OC1=CC=C(C2=CC=C(C(N)=O)C=C2)C=C1)=O)CC3=CC=CC(C4=CC=NC=C4)=C3

InChI

1S/C27H23N3O3/c1-30(18-19-3-2-4-24(17-19)22-13-15-29-16-14-22)27(32)33-25-11-9-21(10-12-25)20-5-7-23(8-6-20)26(28)31/h2-17H,18H2,1H3,(H2,28,31)

InChI key

QTWNORFUQILKJL-UHFFFAOYSA-N

Biochem/physiol Actions

Alpha/beta-hydrolase domain 6 (ABHD6) inhibitor
In mice, WWL70 guards against neuropathic pain stimulated by chronic constriction injury. It decreases the inflammatory response in the ipsilateral spinal cord, dorsal root ganglion (DRG) and sciatic nerve. WWL70 is considered as an anti-inflammatory therapeutic agent, that has the ability to prevent the synthesis of PGE2 (prostaglandin E2) and PGE2-G (PGE2-glyceryl ester).
WWL70 is a selective inhibitor of α/β-hydrolase domain-containing 6 (ABHD6), a serine hydrolase that acts as an alternative hydrolase of the endocannabinoid 2-arachidonoylglycerol (2-AG). It has an IC50 value of 55-70 nM and 90-95% inihibition of ABDH6. WWL70 hs been used in a variety of studies as an ABHD6 antagonist. It was shown to rescue impaired function of mGluR5 signaling, resulting in pain inhibition in arthritic rats. WWL70 was also used to show that ABHD6 is involved in brown adipose function and white adipose browning, and is a potential therapeutic target for obesity and type 2 diabetes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

WWL70 attenuates PGE2 production derived from 2-arachidonoylglycerol in microglia by ABHD6-independent mechanism.
Tanaka M, et al.
Journal of Neuroinflammation, 14(1), 7-7 (2017)
WWL70 protects against chronic constriction injury-induced neuropathic pain in mice by cannabinoid receptor-independent mechanisms.
Wen J, et al.
Journal of Neuroinflammation, 15(1), 9-9 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service