The solubility of the product in DMSO is 20 mg/mL and the solution is clear. Additionally, it exhibits slight solubility in DMSO, ethanol, and methanol.
SML1523
Estetrol
≥98% (HPLC)
Synonym(s):
15α-Hydroxyestriol, 3,15α,16α,17β-Tetrahydroxyestra-1,3,5(10)-triene, E4, Estra-1,3,5(10)-triene-3,15α,16α,17β-tetrol, Oestetrol
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
powder
solubility
DMSO: 20 mg/mL, clear
storage temp.
−20°C
SMILES string
[H][C@@]12CCC3=CC(O)=CC=C3[C@@]1([H])CC[C@@]4(C)[C@@]2([H])[C@@H](O)[C@@H](O)[C@@H]4O
InChI
1S/C18H24O4/c1-18-7-6-12-11-5-3-10(19)8-9(11)2-4-13(12)14(18)15(20)16(21)17(18)22/h3,5,8,12-17,19-22H,2,4,6-7H2,1H3/t12-,13-,14-,15-,16-,17+,18+/m1/s1
InChI key
AJIPIJNNOJSSQC-NYLIRDPKSA-N
General description
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What is the solubility of item SML1523, Estetrol?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service