Skip to Content
Merck
All Photos(1)

Key Documents

SMB00141

Sigma-Aldrich

Garcinoic acid

≥90% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H38O4
CAS Number:
Molecular Weight:
426.59
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

≥90% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

C\C(CC\C=C(/C)C(O)=O)=C/CC\C(C)=C\CC[C@]1(C)CCc2cc(O)cc(C)c2O1

InChI

1S/C27H38O4/c1-19(11-7-13-21(3)26(29)30)9-6-10-20(2)12-8-15-27(5)16-14-23-18-24(28)17-22(4)25(23)31-27/h9,12-13,17-18,28H,6-8,10-11,14-16H2,1-5H3,(H,29,30)/b19-9+,20-12+,21-13+/t27-/m1/s1

InChI key

QOFWRHWADNWKSU-LRXIOGKNSA-N

General description

Natural product derived from plant source.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service