Skip to Content
Merck
All Photos(1)

Key Documents

SMB00102

Sigma-Aldrich

4′,5,7-Trihydroxy-3,6-dimethoxyflavone

≥90% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$348.70

$348.70


Estimated to ship onApril 15, 2025FromSAINT LOUIS


To order products, please contact your local dealer.

Select a Size

Change View
1 MG
$348.70

About This Item

Empirical Formula (Hill Notation):
C17H14O7
CAS Number:
Molecular Weight:
330.29
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

$348.70


Estimated to ship onApril 15, 2025FromSAINT LOUIS


To order products, please contact your local dealer.

Assay

≥90% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

COc1c(O)cc2OC(=C(OC)C(=O)c2c1O)c3ccc(O)cc3

InChI

1S/C17H14O7/c1-22-16-10(19)7-11-12(13(16)20)14(21)17(23-2)15(24-11)8-3-5-9(18)6-4-8/h3-7,18-20H,1-2H3

InChI key

DDNPCXHBFYJXBJ-UHFFFAOYSA-N

General description

Natural product derived from plant source.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F R van Heerden et al.
Fitoterapia, 71(5), 602-604 (2001-07-14)
The major leaf flavonoid of Dodonaea angustifolia, an important South African traditional medicine, has been identified as 5,7,4'-trihydroxy-3,6-dimethoxyflavone (1).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service