Skip to Content
Merck
All Photos(1)

Key Documents

SCP0190

Sigma-Aldrich

MMP 14 (Stomelysin-3) Substrate

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H82N16O17S1
Molecular Weight:
1403.52
UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

Product Name

MMP 14 (Stomelysin-3) Substrate,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥80%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

MCA-Pro-Leu-Ala-Cys-MeOBzl-Trp-Ala-Arg-DNP-Dap-NH2

Application

Matrix metalloproteinases (MMP) are a family of zinc-dependent endopeptidases that degrade matrix proteins. MMPs include collagenases, gelatinases, matrilysins, enamelysins, metalloelastases, stromelysins and other structural protein and receptor lysins. The activity of these enzymes is selectively inhibited or measured by appropriately labeled peptides. MCA-Pro-Leu-Ala-Cys-MeOBzl-Trp-Ala-Arg-DNP-Dap-NH2 is used as a fluorogenic substrate for matrix metalloprotein-14 (Stomelysin, MMP-14).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service