Skip to Content
Merck
All Photos(2)

Documents

SAB5500194

Sigma-Aldrich

Anti-VPAC2 antibody, Rabbit monoclonal

recombinant, expressed in proprietary host, clone SP235, affinity isolated antibody

Synonym(s):

Anti-C16DUPq36.3, Anti-DUP7q36.3, Anti-PACAP-R-3, Anti-PACAP-R3, Anti-VIP-R-2, Anti-VPAC2, Anti-VPAC2R, Anti-VPCAP2R

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in proprietary host

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

SP235, monoclonal

species reactivity

human (tested)

species reactivity (predicted by homology)

rat, mouse

technique(s)

flow cytometry: 1:100
immunohistochemistry: 1:100

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... VIPR2(7434)

General description

Vasoactive intestinal peptide (VIP) receptors 1 and 2, termed VPAC1 and VPAC2, are members of the 7 transmembrane G protein-coupled receptor family and they share homology, but differ in their substrate specificities and expression patterns. Both receptors bind to VIP and pituitary adenylate cyclase-activating polypeptide (PACAP). VPAC2 is upregulated in some carcinoma and lymphoma.

Immunogen

Synthetic peptide derived from human VPAC2 protein.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1 ml rabbit monoclonal antibody purified by protein A/G in PBS/1% BSA buffer pH 7.6 with less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service