Skip to Content
Merck
All Photos(1)

Documents

SAB4300702

Sigma-Aldrich

Anti-TrkB (Ab-705) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-BDNF/NT-3 growth factors receptor precursor, Anti-GP145-TrkB, Anti-GP145-TrkB/GP95-TrkB, Anti-NTRK2, Anti-Trk-B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~145 kDa

species reactivity

mouse, rat, human

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(T-D-Y -Y-R)

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TrkB(4915)

General description

The antibody detects endogenous levels of total TrkB protein.

Immunogen

Peptide sequence around aa. 703-707 (T-D-Y -Y-R), according to the protein NP_001007098.1

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Receptor for brain-derived neurotrophic factor (BDNF), neurotrophin-3 and neurotrophin-4/5 but not nerve growth factor (NGF). Involved in the development and/or maintenance of the nervous system. This is a tyrosine-protein kinase receptor. Known substrates for the TRK receptors are SHC1, PI-3 kinase, and PLC-gamma-1.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elahe Karimi et al.
Drug design, development and therapy, 13, 2513-2532 (2019-08-24)
Nowadays, the drug delivery system is important in the treatment of diseases. A polymeric nanoparticle modified by oleic acid (NPMO) as a Thymol (Thy) drug release system was synthesized from Thymbra spicata and its neurotrophic and angiogenic effects on rat's
Sabah Shiri et al.
RSC advances, 9(67), 38912-38927 (2019-11-27)
Drug delivery systems have been of interest to researchers. The effects of synthesized nano-polymers as silibinin and silymarin extract drug delivery systems on olfactory ensheathing cells under normal and high-glucose conditions were studied. The structure of the nanopolymer was characterized
Arjun Singh et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 34(4), e14099-e14099 (2021-02-25)
Data on the neuromodulatory effects of brain-derived neurotrophic factor (BDNF) in the gastrointestinal tract were recently reported, but there are still no data on the presence, distribution, and release of BDNF in the gastrointestinal tract, including the internal anal sphincter

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service