Skip to Content
Merck
All Photos(1)

Key Documents

S6433

Sigma-Aldrich

Anti-STAT6 (787-804) antibody produced in rabbit

whole antiserum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

mol wt

antigen 100 kDa

contains

0.035% sodium azide

species reactivity

mouse (weakly), human

technique(s)

western blot (chemiluminescent): 1:1,000 using a human A431 cell lysate

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STAT6(6778)
mouse ... Stat6(20852)

Specificity

The antibody is specific for human STAT6. The antibody also reacts weakly with mouse STAT6. This antiserum is not recommended for immunoprecipitation.

Immunogen

synthetic peptide corresponding to human STAT6 (amino acids 787-804) conjugated to KLH.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service