Skip to Content
Merck
All Photos(1)

Key Documents

S0198

Sigma-Aldrich

Monoclonal Anti-Sonic Hedgehog peptide (N-terminal) antibody produced in rat

clone 171001, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-Dsh, Anti-Hhg1, Anti-Hx, Anti-Shh

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.41

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

171001, monoclonal

form

lyophilized powder

species reactivity

mouse, human

technique(s)

western blot: 1-2 μg/mL

isotype

IgG2a

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SHH(6469)
mouse ... Shh(20423)

Immunogen

purified, E. coli-derived recombinant mouse 6× histidine-tagged sonic hedgehog N-terminal peptide (aa 25-198).

Biochem/physiol Actions

Shh protein is secreted by Purkinje cells in the cerebellum with both autocrine and paracrine functions. The Shh signaling is mediated by the interaction of Shh with transmembrane proteins, Patched and Smoothened. The Shh pathway cross-talks Notch and Wnt pathways and acts via downstream targets such as cyclins, gamma catenin, myc and other transcription factors. The Hh family members have multiple roles such as morphogens, cell survival factors, setting anterior-posterior axis and digit formation during the development of brain and nervous system.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessia Omenetti et al.
American journal of physiology. Gastrointestinal and liver physiology, 294(3), G595-G598 (2008-01-26)
Hedgehog (Hh) signaling modulates tissue remodeling by controlling the fate of Hh-responsive cells. Healthy adult livers exhibit little Hh activity. However, cells involved in adult liver repair, including myofibroblasts and progenitors, are capable of producing and responding to Hh ligands.
Shohreh F Farzan et al.
American journal of physiology. Gastrointestinal and liver physiology, 294(4), G844-G849 (2008-02-02)
The Hedgehog (Hh) family of secreted proteins is necessary for aspects of the development and maintenance of the gastrointestinal tract. Hh is thought to function as a morphogen, a mitogen, a cell survival factor, and an axon guidance factor. Given
Fergal C Kelleher
Carcinogenesis, 32(4), 445-451 (2010-12-28)
To conduct a systematic review of the role that the hedgehog signaling pathway has in pancreatic cancer tumorigenesis. PubMed search (2000-2010) and literature based references. Firstly, in 2009 a genetic analysis of pancreatic cancers found that a core set of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service