Skip to Content
Merck
All Photos(1)

Key Documents

PZ0367

Sigma-Aldrich

PF-02413873

≥98% (HPLC)

Synonym(s):

4-[3-Cyclopropyl-1-(mesylmethyl)-5-methyl-1H-pyrazol-4-yl]oxy-2,6-dimethylbenzonitrile, 4-[[3-Cyclopropyl-5-methyl-1-[(methylsulfonyl)methyl]-1H-pyrazol-4-yl]oxy]-2,6-dimethyl-benzonitrile, PF 2413873, PF-2413873

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21N3O3S
CAS Number:
Molecular Weight:
359.44
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

SMILES string

CC1=C(C#N)C(C)=CC(OC2=C(C)N(CS(=O)(C)=O)N=C2C3CC3)=C1

InChI

1S/C18H21N3O3S/c1-11-7-15(8-12(2)16(11)9-19)24-18-13(3)21(10-25(4,22)23)20-17(18)14-5-6-14/h7-8,14H,5-6,10H2,1-4H3

InChI key

QSFGZNVRVZHUGV-UHFFFAOYSA-N

Biochem/physiol Actions

PF-02413873 is a potent, selective and fully competitive non-steroidal progesterone receptor (PR) antagonist that blocks progesterone binding and PR nuclear translocation. PF-02413873 suppresses endometrial growth in the macaque and human.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter J Bungay et al.
Drug metabolism and disposition: the biological fate of chemicals, 39(8), 1396-1405 (2011-05-06)
The recently discovered selective nonsteroidal progesterone receptor (PR) antagonist 4-[3-cyclopropyl-1-(methylsulfonylmethyl)-5-methyl-1H-pyrazol-4-yl]oxy-2,6-dimethylbenzonitrile (PF-02413873) was characterized in metabolism studies in vitro, in preclinical pharmacokinetics in rat and dog, and in an initial pharmacokinetic study in human volunteers. Clearance (CL) of PF-02413873 was found
David C Howe et al.
The Journal of pharmacology and experimental therapeutics, 339(2), 642-653 (2011-08-19)
There is considerable ongoing investment in the research and development of selective progesterone receptor (PR) modulators for the treatment of gynecological conditions such as endometriosis. Here, we provide the first report on the clinical evaluation of a nonsteroidal progesterone receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service