Skip to Content
Merck
All Photos(1)

Key Documents

H9538

Sigma-Aldrich

HNE-DMA

hexane solution, ≥85% (GC)

Synonym(s):

(E)-4-Hydroxynonenal-dimethylacetal, 4-HNE-DMA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H22O3
Molecular Weight:
202.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥85% (GC)

form

hexane solution

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCC(O)\C=C\C(OC)OC

InChI

1S/C11H22O3/c1-4-5-6-7-10(12)8-9-11(13-2)14-3/h8-12H,4-7H2,1-3H3/b9-8+

InChI key

ATGIHMSJAARJTF-CMDGGOBGSA-N

Biochem/physiol Actions

Stable form of HNE; toxic second messenger of free radicals.

Preparation Note

To prepare HNE from HNE-DMA: evaporate the hexane under a gentle stream of nitrogen at room temperature. Working at 4 °C, add 1 ml (5 mg size) or 0.4 ml (2 mg size) of 1 mM cold HCl and stir for 45 minutes, or alternatively, close the vial and shake for 30 minutes. Upon hydrolysis, the initially cloudy solution becomes almost clear. This solution now has a pH of ∼3, contains ∼5 mg (5 mg size) or ∼2 mg (2 mg size) of the aldehyde (HNE), and is stable for at least 24 hours if kept at 4 °C. 6.8 mg HNE-DMA in 1 mL of hexane solution, giving 5 mg of HNE after hydrolysis.It is recommended that the freshly prepared HNE solution be used immediately.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Héléna Alamil et al.
Toxicology letters, 331, 57-64 (2020-05-23)
Electrophilic compounds present in humans, originating from endogenous processes or pollutant exposures, pose a risk to health though their reaction with nucleophilic sites in protein and DNA. Among this chemical class, aldehydes are mainly present in indoor air and they
Femke van Diggelen et al.
PloS one, 14(3), e0213663-e0213663 (2019-03-23)
Parkinson's Disease (PD) is a neurodegenerative disease for which there currently is no cure. Aggregation of the pre-synaptic protein α-synuclein (aSN) into oligomers (αSOs) is believed to play a key role in PD pathology, but little is known about αSO
Thomas Van Hecke et al.
Foods (Basel, Switzerland), 10(11) (2021-11-28)
Oxidative reactions during cooking and gastrointestinal digestion of meat and fish lead to the formation of various lipid- and protein oxidation products, some of which are toxic. In the present study, it was investigated how the addition of 3% butter
Mutian Jia et al.
Nature immunology, 21(7), 727-735 (2020-06-17)
Stimulator-of-interferon genes (STING) is vital for sensing cytosolic DNA and initiating innate immune responses against microbial infection and tumors. Redox homeostasis is the balance of oxidative and reducing reactions present in all living systems. Yet, how the intracellular redox state
Wejdane El Manaa et al.
Autophagy, 17(12), 4363-4385 (2021-05-26)
Parkinson disease (PD)-affected brains show consistent endoplasmic reticulum (ER) stress and mitophagic dysfunctions. The mechanisms underlying these perturbations and how they are directly linked remain a matter of questions. XBP1 is a transcription factor activated upon ER stress after unconventional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service