Skip to Content
Merck
All Photos(2)

Key Documents

GBL246877

Grace Bio-Labs silicone gasket for ProPlate® microarray system

fits 16 wells, diam. 6 mm

Synonym(s):

microarray chambers for slides, silicone slide gasket

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122600
NACRES:
NB.15

sterility

non-sterile

feature

wells 16

packaging

pack of 10 ea

manufacturer/tradename

Grace Bio-Labs 246877

diam.

6 mm

size

16 wells

color

none

Looking for similar products? Visit Product Comparison Guide

General description

Replacement Silicone Gaskets for Grace BioLabs Proplate slide modules.

While the silicone gaskets affixed to wells may be re-used multiple times, their lifespan may be shortened by washing conditions, repeated autoclaving and prolonged exposure to harsher types of chemicals. Therefore, replacement gaskets are available and may be easily positioned and applied to the chambers after simply peeling the old gasket off and throwing it away.

Application

  • Protein and peptides microarrays
  • cDNA and oligonucleotides microarrays
  • Proteomics
  • Protein expression analysis
  • Antibody profiling

Legal Information

ProPlate is a registered trademark of Grace Bio-Labs, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service