EP952010069
Eppendorf® Uvette® routine pack, reclosable box
220 nm – 1,600 nm, 50 – 2,000 µL, pack of 200 ea
All Photos(1)
About This Item
UNSPSC Code:
41121813
Recommended Products
material
(UV-transparent plastic)
packaging
pack of 200 ea
manufacturer/tradename
Eppendorf® 952010069
capacity
50 -– 2,000 μL
General description
UVette® routine pack 220 nm – 1,600 nm, reclosable box, Eppendorf Quality, 50 – 2,000 µL, 200 pcs.
- UVettes in routine packs are ideal for routine applications.
- These packs allow a safe storage of the UVettes in a reclosable box, while offering convenient access to each cuvette for easy filling.
- UV transparent plastic, free of fluoropolymers and other halogenated hydrocarbons
- Two built-in optical path lengths in a single cuvette - just turn the UVette 90° to change from 10 mm and 2 mm
- Volume markings for 500 µL and 1,000 µL
- Deep-lying optical window prevents scratches
- Tapered cuvette base for optimal filling, frosted grip for labeling
- Ideal for use in the Eppendorf BioPhotometer and Eppendorf BioSpectrometer, adapters available for use with other common spectrophotometers
- Routine pack for convenient access to each UVette and safe storage in a reclosable box
Features and Benefits
- UVettes in routine packs are ideal for routine applications.
- These packs allow a safe storage of the UVettes in a reclosable box, while offering convenient access to each cuvette for easy filling.
- UV transparent plastic, free of fluoropolymers and other halogenated hydrocarbons
- Two built-in optical path lengths in a single cuvette - just turn the UVette 90° to change from 10 mm and 2 mm
- Volume markings for 500 µL and 1,000 µL
- Deep-lying optical window prevents scratches
- Tapered cuvette base for optimal filling, frosted grip for labeling
- Ideal for use in the Eppendorf BioPhotometer and Eppendorf BioSpectrometer, adapters available for use with other common spectrophotometers
- Routine pack for convenient access to each UVette and safe storage in a reclosable box
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
UVette is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)