Skip to Content
Merck
All Photos(1)

Key Documents

EP4982000411

Eppendorf® Multipette® M4

new

Includes holder for wall mount or pipette carousel

Synonym(s):

Eppendorf® Multipette® M4, pipettors

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121506

feature

adjustable
manual
no. of channels single
Includes holder for wall mount or pipette carousel

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 4982000411

volume

1 µl to 50 mL
1 μL , Adjustable in 1µL increments

Looking for similar products? Visit Product Comparison Guide

General description

Multipette M4 basic including holder (for wall mount and/or pipette carousel)
With the Multipette M4/Combitip System, volumes are dispensed using the positive displacement principle. The liquid is directly dispensed without an air cushion, ensuring the correct volume is always dispensed regardless of the density, viscosity and volatility of the liquid. The Multipette M4 offers a unique volume range of 1 μL – 10 mL, and is the only mechanical dispenser that dispenses the aspirated liquid up to 100 times without refilling the tip. The innovative step counter supports error-free working even after an interruption or distraction during the dispensing procedure. A fully emptied Combitip can be easily ejected with one hand using the operation lever without directly touching the tip.

Features and Benefits

1 µL – 10 mL

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Multipette is a registered trademark of Eppendorf AG

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service