EP3125000010
Eppendorf Research® Plus G
8-channel, variable, volume (0.5-10 μL), medium gray
Synonym(s):
Adjustable volume pipette
About This Item
Recommended Products
feature
8-channel, variable
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 3125000010
volume
(0.5-10 μL)
× 10 μL
color
medium gray
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Product features
It is a good feeling to know you are doing the right thing – to know you are working with one of the most advanced pipettes in the world. A spring loaded tip cone, a secondary calibration option, an improved volume display – and all that in an ultra light, fully autoclavable pipette: That is the Eppendorf Research plus pipette.
Accoladed product
The Research plus mechanical pipette was accoladed with the Bronze “Seal of Quality”. These seals are given to the top 0.1 % of products used in laboratories that have continually received the highest user review ratings. The award is praised as providing “invaluable” insights for scientists and intends to help them choose the ideal laboratory product for their working environment.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service