Skip to Content
Merck
All Photos(1)

Key Documents

EHU035131

Sigma-Aldrich

MISSION® esiRNA

targeting human KLF5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCACCTGAGAACTGGCCTCTACAAATCCCAGAGACCGTGCGTAACACACATCAAGACAGAACCTGTTGCCATTTTCAGCCACCAGAGTGAAACGACTGCCCCTCCTCCGGCCCCGACCCAGGCCCTCCCTGAGTTCACCAGTATATTCAGCTCACACCAGACCGCAGCTCCAGAGGTGAACAATATTTTCATCAAACAAGAACTTCCTACACCAGATCTTCATCTTTCTGTCCCTACCCAGCAGGGCCACCTGTACCAGCTACTGAATACACCGGATCTAGATATGCCCAGTTCTACAAATCAGACAGCAGCAATGGACACTCTTAATGTTTCTATGTCAGCTGCCATGGCAGGCCTTAACACACACACCTCTGCTGTTCCGCAGACTGCAGTGAAACAATTCCAGGGCAT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Chen et al.
European review for medical and pharmacological sciences, 24(8), 4224-4231 (2020-05-07)
This study aims to investigate the expression characteristics of Krüppel-like factor 5 (KLF5) in gastric cancer (GC) and its potential correlation to pathological indexes in GC patients. Molecular mechanisms underlying the regulatory effect of KLF5 on GC progression are explored.
Qiu-Yu Wang et al.
Molecular oncology, 14(9), 2203-2230 (2020-05-28)
Long noncoding RNAs (lncRNAs) have important regulatory roles in cancer biology. Although some lncRNAs have well-characterized functions, the vast majority of this class of molecules remains functionally uncharacterized. To systematically pinpoint functional lncRNAs, a computational approach was proposed for identification
Kaixin Wangzhou et al.
Frontiers in physiology, 11, 606967-606967 (2021-02-20)
Human periodontal ligament cells (hPDLCs) play a vital role in cell regeneration and tissue repair with multi-directional differentiation potential. microRNAs (miRs) are implicated in the osteogenesis of hPDLCs. This study explored the mechanism of miR-143-3p in osteogenesis of hPDLCs. Osteogenic
Liyuan Gao et al.
Life sciences, 264, 118696-118696 (2020-11-07)
Liver fibrosis is a difficult problem in the medical field. We previously reported that curcumol, a bioactive substance, may inhibit the pathological angiogenesis of liver sinusoidal endothelial cells (LSECs) and play a good anti-hepatic fibrosis effect. However, the mechanism of
Yubo Liu et al.
Nature communications, 11(1), 5898-5898 (2020-11-21)
O-GlcNAc modification plays critical roles in regulating the stress response program and cellular homeostasis. However, systematic and multi-omics studies on the O-GlcNAc regulated mechanism have been limited. Here, comprehensive data are obtained by a chemical reporter-based method to survey O-GlcNAc

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service