Key Documents
CLS3479
Corning® NETWELL® 6 well plates
clear , polyester membrane, round wells, clear wells, polyester plate, sterile, pkg of 48 (8 x 6 well inserts)
Synonym(s):
cell culture inserts, plate inserts, tissue culture dishes, tissue culture plates
About This Item
UNSPSC Code:
41122107
NACRES:
NB.13
Product Name
Corning® Netwell® inserts, membrane diam. 24 mm, membrane size 74 μm , mesh
material
clear
clear wells
polyester membrane
polyester plate
round wells
sterility
sterile
feature
lid
skirt
packaging
pkg of 48 (8 x 6 well inserts)
manufacturer/tradename
Corning 3479
membrane diam.
24 mm
membrane size
74 μm , mesh
size
6 wells , per insert
General description
Netwells are polystyrene inserts fitted with polyester mesh bottoms that function as tissue carriers and strainers for:
  • Immunocytochemical staining
  • Explant culture
  • 3-D tissue culture
  • Coarse filtration of cell suspension
Features and Benefits
  • Corning® Netwell® inserts are 24mm diameter netwell inserts have 74μm polyester mesh bottoms attached to polystyrene to rings or housing.
  • Supplied sterile and pre-loaded in 6-well microplates.
  • 24mm Netwell inserts fit in Corning 50 mL plastic centrifuge tubes.
Legal Information
Corning is a registered trademark of Corning, Inc.
NETWELL is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)