CLS11425
Corning® Erlenmeyer flask, with aseptic connector
volume 500 mL, I.D. 1/8 in., sterile
Synonym(s):
Corning container
About This Item
Recommended Products
material
flat bottom flask
polycarbonate
polypropylene cap
description
plain
sterility
sterile
feature
cap: no
C-Flex tubing
packaging
case of 2
manufacturer/tradename
Corning 11425
I.D.
1/8 in.
capacity
500 mL
volume
500 mL
fitting
male luer lock
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)