CLS1015P1L
Corning® reusable beaker with handle and spout
polypropylene, size 1 L
Synonym(s):
Corning container
About This Item
Recommended Products
material
polypropylene
feature
graduations (10 mL)
manufacturer/tradename
Corning 1015P-1L
H × O.D.
167 mm × 125 mm
size
1 L
Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service