COLO 792
93052616, human skin (brain metastasis), Epithelial
Synonym(s):
COLO792 Cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
Product Name
COLO 792, 93052616
biological source
human skin (brain metastasis)
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
metastasis
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human malignant melanoma
Cell Line Description
Derived from a brain metastasis of a 62 year old male with melanoma. The cells bleb during culture.
Application
Tumourigenicity studies
DNA Profile
STR-PCR Data: Amelogenin: X,Y
CSF1PO: 11
D13S317: 11,12
D16S539: 12
D5S818: 10,12
D7S820: 10,11
THO1: 6,9
TPOX: 8,11
vWA: 16,17
CSF1PO: 11
D13S317: 11,12
D16S539: 12
D5S818: 10,12
D7S820: 10,11
THO1: 6,9
TPOX: 8,11
vWA: 16,17
Culture Medium
RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service