Skip to Content
Merck
All Photos(1)

Key Documents

3T3 Swiss Albino

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 85022108, mouse embryo, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

mouse embryo

packaging

tube of 5 μg 85022108-DNA-5UG
pkg of vial of cells 85022108-1VL

growth mode

Adherent

karyotype

2n = 40; hypertriploid

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

General description

Product has lost contact inhibition

Cell Line Origin

Mouse Swiss Albino embryo fibroblast

Cell Line Description

Evolved under conditions favouring retention of the property of contact inhibition of cell proliferation; they exhibit much more contact inhibition than diploid fibroblasts freshly plated in culture. Readily transformed by polyoma and SV40 viruses resulting in reduced sensitivity to contact inhibition. Cells have now lost their contact inhibition.

Application

Contact inhibition and virus studies: polyoma and SV40 viruses

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:10 to 1:20 i.e. seeding at 5x1,000 to 1x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service