B932-05
Bovine Endometrial Epithelial Cells: BEnEpC (Cryovial)
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
biological source
bovine endometrium
packaging
pkg of 500,000 cells
manufacturer/tradename
Cell Applications, Inc
growth mode
Adherent
karyotype
2n = 60
morphology
epithelial
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
infectious diseases
shipped in
dry ice
storage temp.
−196°C
General description
Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.
BEnEpC provide an excellent model system to study many aspects of epithelial function and disease, especially those related to the endometrium and pregnancy-related signaling, such as seminal plasma induced prostaglandins F2α, (PGF2α), E2 (PGE2) and interleukin-6 release by endometrial cells (Norrby, 2012; Madej, 2012; Malgorzata, 2013), or the role of PLA in mediating oxytocin-stimulated production of prostaglandins (Tithof, 2007). BEnEpC have also been used to demonstrate the association between variants of gap junctions related genes and conception rate (Sugimoto, 2013), and to investigate cellular adherence and cytotoxicity of Histophilus somni (Hoshinoo, 2009).
BEnEpC provide an excellent model system to study many aspects of epithelial function and disease, especially those related to the endometrium and pregnancy-related signaling, such as seminal plasma induced prostaglandins F2α, (PGF2α), E2 (PGE2) and interleukin-6 release by endometrial cells (Norrby, 2012; Madej, 2012; Malgorzata, 2013), or the role of PLA in mediating oxytocin-stimulated production of prostaglandins (Tithof, 2007). BEnEpC have also been used to demonstrate the association between variants of gap junctions related genes and conception rate (Sugimoto, 2013), and to investigate cellular adherence and cytotoxicity of Histophilus somni (Hoshinoo, 2009).
Cell Line Origin
Uterus
Application
endometrial function, growth factor production, conception rate, cell adherence, uterine lining, maintenance of uterine patency, uterine contraction
Components
Bovine Endometrial Basal Media containing 10% FBS & 10% DMSO
Preparation Note
- 2nd passage, >500,000 cells in Bovine Endometrial Basal Media containing 10% FBS & 10% DMSO
- Can be cultured at least 10 doublings
Subculture Routine
Please refer to the BEnEpC Culture Protocol.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service