89050907
LLC-RK1 cell line
rabbit kidney, Epithelial-like
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
NACRES:
NA.81
Recommended Products
product name
LLC-RK1 cell line, from rabbit kidney
biological source
rabbit kidney
form
liquid
growth mode
Adherent
karyotype
Modal no. 45 (2n=46)
morphology
Epithelial-like
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Description
Derived from kidneys taken from New Zealand white rabbits. The cells are susceptible to a range of viruses including rubella, vaccinia and herpes. Rubella replicates to a high titre.
Culture Medium
Medium 199 + 2 mM Glutamine + 10% Fetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 5x10,000 to 3x100,000 cells/cm2using 0.25% trypsin; 5% CO2; 37°C. NB: this line is extremely sensitive to trypsin.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
DEVELOPMENT AND CHARACTERISTICS OF THE RABBIT KIDNEY CELL STRAIN, LLC-RK.
R N HULL et al.
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.), 118, 1054-1059 (1965-04-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service