NISTRM8568
Nitrogen and oxygen isotopes in nitrate
NIST® RM 8568
Synonym(s):
Potassium nitrate
About This Item
Recommended Products
grade
certified reference material
Quality Level
isotopic purity
15N
17O
18O
manufacturer/tradename
NIST®
mp
334 °C (lit.)
format
mixture
SMILES string
[K+].[O-][N+]([O-])=O
InChI
1S/K.NO3/c;2-1(3)4/q+1;-1
InChI key
FGIUAXJPYTZDNR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
Nitrogen (15N), Oxygen (17O/18O)
See certificate for values and more details at nist.gov/SRM.
Legal Information
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Ox. Sol. 3
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service