Skip to Content
Merck
All Photos(1)

Documents

LIGHTOX17

Sigma-Aldrich

LightOx17

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H27NO2
CAS Number:
Molecular Weight:
373.49
UNSPSC Code:
12352101
NACRES:
NA.22

Assay

≥98% (HPLC)

form

powder

storage temp.

2-8°C

Application

Fluorescence microscopy, fluorescence spectroscopy.

Features and Benefits

LightOx17 is a fluorescent molecule that exhibits solvatochromatic fluorescence emission when excited with UV-A and violet (405 nm) light sources. In a cellular environment it exhibits predominately mitochrondrial localisation and, when activated by UV-A or 405 nm absorption wavelengths, elicits the release of reactive oxygen species (ROS) for controlled, localised cell death.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Other Notes

Chemical Equivalency 1ea=1mg

Legal Information

LightOx is a trademark of LightOx Ltd.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Fluorescent retinoic acid analogues for use in cellular imaging, protein-ligand binding assays, and flow cytometry to bypass traditional problems associated with fluorescent probes.

Fluorescent retinoic acid analogues for use in cellular imaging, protein-ligand binding assays, and flow cytometry to bypass traditional problems associated with fluorescent probes.

Fluorescent retinoic acid analogues for use in cellular imaging, protein-ligand binding assays, and flow cytometry to bypass traditional problems associated with fluorescent probes.

Fluorescent retinoic acid analogues for use in cellular imaging, protein-ligand binding assays, and flow cytometry to bypass traditional problems associated with fluorescent probes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service