ERMFD304
Silica, colloidal
ERM®, certified reference material
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
85151701
NACRES:
NA.24
Recommended Products
grade
certified reference material
Agency
ERM®
form
particles
manufacturer/tradename
JRC
format
neat
Looking for similar products? Visit Product Comparison Guide
Analysis Note
For more information please see:
ERMFD304
ERMFD304
Legal Information
ERM is a registered trademark of European Commission
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Margarida Bastos et al.
Biophysical journal, 94(6), 2128-2141 (2007-11-23)
The energetics and partition of two hybrid peptides of cecropin A and melittin (CA(1-8)M(1-18) and CA(1-7)M(2-9)) with liposomes of different composition were studied by time-resolved fluorescence spectroscopy, isothermal titration calorimetry, and surface plasmon resonance. The study was carried out with
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service