Skip to Content
Merck
All Photos(1)

Key Documents

936685

Sigma-Aldrich

Grubbs Catalyst® M102 ChemBeads

Synonym(s):

Grubbs Catalyst® M102, Benzylidene-bis(tricyclohexylphosphine)dichlororuthenium, Bis(tricyclohexylphosphine)benzylidine ruthenium(IV) dichloride ChemBeads, Dichloro(benzylidene)bis(tricyclohexylphosphine)ruthenium(II) ChemBeads, Grubbs Catalyst® 1st Generation ChemBeads, Grubbs Catalyst® M1a (C823) ChemBeads

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H72Cl2P2Ru
CAS Number:
Molecular Weight:
822.96
MDL number:
UNSPSC Code:
12352103

form

solid ()

Quality Level

composition

4-6 wt% loading of catalyst

reaction suitability

reagent type: catalyst
core: ruthenium
reaction type: Ring-Opening Polymerization

mp

153 °C (dec.)

SMILES string

Cl[Ru]([P](C1CCCCC1)(C2CCCCC2)C3CCCCC3)([P](C4CCCCC4)(C5CCCCC5)C6CCCCC6)(Cl)=CC7=CC=CC=C7
C[Ph].P(C1CCCCC1)(C2CCCCC2)C3CCCCC3.P(C4CCCCC4)(C5CCCCC5)C6CCCCC6.Cl.Cl.[Ru]

InChI

1S/2C18H33P.C7H6.2ClH.Ru/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-7-5-3-2-4-6-7;;;/h2*16-18H,1-15H2;1-6H;2*1H;

InChI key

AULHKSXXVKDNBT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

ChemBeads glass beads coated with Grubbs Catalyst® M102.

Application

First metathesis catalyst to be widely used in organic synthesis. Useful for acyclic diene metathesis polymerization (ADMET), Ring-Opening Metathesis Polymerization (ROMP) of strained cyclic olefins, ring opening metathesis (ROM), olefin cross metathesis (CM) and ring closing metathesis (RCM) of terminal olefins under a variety of reactions conditions.
ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.
For larger scale uses, product also available in powdered form (579726)

Legal Information

Grubbs Catalyst 7578
Grubbs Catalyst is a registered trademark of Umicore AG & Co. KG

related product

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongwon Lee et al.
International journal of nanomedicine, 3(4), 471-476 (2008-01-01)
The overproduction of hydrogen peroxide is implicated in the progress of numerous life-threatening diseases and there is a great need for the development of contrast agents that can detect hydrogen peroxide in vivo. In this communication, we present a new
Thomas A. Kirkland et al.
The Journal of organic chemistry, 62(21), 7310-7318 (2001-10-24)
Ruthenium alkylidene 1 and molybdenum alkylidene 2 have been utilized in the ring-closing metathesis (RCM) of dienes containing gem-disubstituted olefins to yield tri- and tetrasubstituted cyclic olefins. Dienes with sterically demanding and/or electron-withdrawing substituents such as Ph, CO(2)Me, and (t)()Bu

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service