Skip to Content
Merck
All Photos(2)

Key Documents

70167

Millipore

Malt Extract for Biotechnology and Microbiology

for Biotechnology and Microbiology

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.74
Pricing and availability is not currently available.

Product Name

Malt extract, suitable for microbiology,

Purified and clarified malt extract, supplemented with growth factors

biological source

corn

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

chlorides, ~2%
reducing sugars (as maltose), ≥60%

ign. residue

~3%

loss

~6% loss on drying

pH

5-6 (3% in H2O)

solubility

H2O: 3%, faintly turbid, yellow

application(s)

microbiology

Looking for similar products? Visit Product Comparison Guide

General description

Malt extract powder is prepared from an aqueous extract of sprouted malt grains and dried at low temperatures to preserve nutrients. This malt extract is a purified and clarified malt extract, supplemented with growth factors. It is easily soluble in water. It contains a mix of carbohydrates (mainly maltose) and growth factors. It is ideally suitable for use in media employed for cultivation of fungi.

Application

Malt extract powder is added to specific culture media to enhance the nutrient quality of the media. It is most suited to enhance the growth and cultivation of fungi, yeasts, and molds

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Losee L Ling et al.
Nature, 517(7535), 455-459 (2015-01-07)
Antibiotic resistance is spreading faster than the introduction of new compounds into clinical practice, causing a public health crisis. Most antibiotics were produced by screening soil microorganisms, but this limited resource of cultivable bacteria was overmined by the 1960s. Synthetic

Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service