Skip to Content
Merck
All Photos(1)

Key Documents

28-5009

Sigma-Aldrich

Starch solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12000000

form

liquid

availability

available only in Japan

concentration

~1 % (w/v)

dilution

(for analytical testing)

Looking for similar products? Visit Product Comparison Guide

Suitability

for titration

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service