Skip to Content
Merck
All Photos(1)

Key Documents

09-1287

Sigma-Aldrich

Ethylenediamine

SAJ special grade, ≥99.0%

Synonym(s):

1,2-Diaminoethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2NH2
CAS Number:
Molecular Weight:
60.10
Beilstein:
605263
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

grade

SAJ special grade

vapor density

2.07 (vs air)

vapor pressure

10 mmHg ( 20 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

716 °F

expl. lim.

16 %

availability

available only in Japan

refractive index

n20/D 1.4565 (lit.)

pH

12.2 (20 °C, 110 g/L)

bp

118 °C (lit.)

mp

8.5 °C (lit.)

density

0.899 g/mL at 25 °C (lit.)

SMILES string

NCCN

InChI

1S/C2H8N2/c3-1-2-4/h1-4H2

InChI key

PIICEJLVQHRZGT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Resp. Sens. 1B - Skin Corr. 1B - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Zuidema
Pharmaceutisch weekblad. Scientific edition, 7(4), 134-140 (1985-08-23)
Ethylenediamine is an excipient with many industrial and pharmaceutical uses. It is included in creams as a stabilizer and in aminophylline as the counter ion of theophylline. Ethylenediamine is one of the most frequent contact sensitizers, producing local and generalized
Po Ying Yeh et al.
Langmuir : the ACS journal of surfaces and colloids, 28(46), 16227-16236 (2012-11-01)
We report a novel nonfouling passivation method using poly(ethylene glycol) (PEG) engraftment on the surfaces of poly(dimethylsiloxane) (PDMS) microfluidic devices sealed with SU-8. To achieve bonding between the PDMS and SU-8 surfaces, the PDMS surface was first functionalized with amines
Ana Carolina M S Dias et al.
Biosensors & bioelectronics, 44, 216-221 (2013-02-23)
An immunosensor for the non-structural protein 1 (NS1) of the dengue virus based on carbon nanotube-screen printed electrodes (CNT-SPE) was successfully developed. A homogeneous mixture containing carboxylated carbon nanotubes was dispersed in carbon ink to prepare a screen printed working
Changhai Xu et al.
Carbohydrate polymers, 92(1), 249-253 (2012-12-12)
There exists a misunderstanding on the TAED-activated peroxide system in the textile industry that H(2)O(2) used in excess of the stoichiometric amount could produce an addition effect on bleaching of cotton under alkaline conditions. In this study, a critical reinvestigation
Kavita Sablok et al.
Journal of hazardous materials, 248-249, 322-328 (2013-02-19)
Binding of electron-deficient trinitrotoluene (TNT) to the electron rich amine groups on a substrate form specific charge-transfer Jackson-Meisenheimer (JM) complex. In the present work, we report formation of specific JM complex on amine functionalized reduced graphene oxide/carbon nanotubes- (a-rGO/CNT) nanocomposite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service