Skip to Content
Merck
All Photos(1)

Key Documents

07-5360

Sigma-Aldrich

1,4-Dioxane

SAJ first grade

Synonym(s):

Diethylene oxide, Dioxane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
Beilstein:
102551
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

SAJ first grade

vapor density

3 (vs air)

vapor pressure

27 mmHg ( 20 °C)
40 mmHg ( 25 °C)

Assay

>99.0% (GC)

form

liquid

autoignition temp.

356 °F

expl. lim.

22 %

availability

available only in Japan

refractive index

n20/D 1.422 (lit.)

pH

6.0-8 (20 °C, 500 g/L)

bp

100-102 °C (lit.)

mp

10-12 °C (lit.)

density

1.034 g/mL at 25 °C (lit.)

SMILES string

C1COCCO1

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2

InChI key

RYHBNJHYFVUHQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

recommended

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huijiao Wang et al.
Journal of hazardous materials, 294, 90-98 (2015-04-12)
Degradation of 1,4-dioxane by ozonation, electrolysis, and their combined electro-peroxone (E-peroxone) process was investigated. The E-peroxone process used a carbon-polytetrafluorethylene cathode to electrocatalytically convert O2 in the sparged ozone generator effluent (O2 and O3 gas mixture) to H2O2. The electro-generated
Xiaohua Liu et al.
Biomaterials, 31(2), 259-269 (2009-09-29)
It remains a challenge to synthesize functional materials that can develop advanced scaffolding architectures for tissue engineering. In this study, a series of biodegradable amphiphilic poly(hydroxyalkyl (meth)acrylate)-graft-poly(l-lactic acid) (PHAA-g-PLLA) copolymers have been synthesized and fabricated into nano-fibrous scaffolds. These copolymers
Joshua J Pacheco et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(23), 8363-8367 (2014-06-10)
Terephthalic acid (PTA), a monomer in the synthesis of polyethylene terephthalate (PET), is obtained by the oxidation of petroleum-derived p-xylene. There is significant interest in the synthesis of renewable, biomass-derived PTA. Here, routes to PTA starting from oxidized products of
Christof Aellig et al.
ChemSusChem, 5(9), 1737-1742 (2012-07-05)
The dehydration of D-fructose to 5-hydroxymethylfurfural was studied under single-phase conditions in the low boiling solvent 1,4-dioxane at moderate temperatures in the presence of the solid acid-catalyst Amberlyst-15. The reaction was first examined and optimized under batch conditions, where it
Alkene trifluoromethylation coupled with C-C bond formation: construction of trifluoromethylated carbocycles and heterocycles.
Hiromichi Egami et al.
Angewandte Chemie (International ed. in English), 52(14), 4000-4003 (2013-02-27)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service