05-2660
Chloral hydrate
SAJ first grade, ≥99.5%
Synonym(s):
Trichloroacetaldehyde hydrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Cl3CCH(OH)2
CAS Number:
Molecular Weight:
165.40
Beilstein:
1698497
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
grade
SAJ first grade
Assay
≥99.5%
availability
available only in Japan
mp
57 °C (lit.)
SMILES string
OC(O)C(Cl)(Cl)Cl
InChI
1S/C2H3Cl3O2/c3-2(4,5)1(6)7/h1,6-7H
InChI key
RNFNDJAIBTYOQL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Virginie Bros-Facer et al.
Human molecular genetics, 23(16), 4187-4200 (2014-03-29)
Amyotrophic lateral sclerosis (ALS) is a fatal, neurodegenerative disorder in which motor neurons in the spinal cord and motor cortex degenerate. Although the majority of ALS cases are sporadic, mutations in Cu-Zn superoxide dismutase-1 (SOD1) are causative for 10-20% of
Simone Jürschik et al.
Journal of mass spectrometry : JMS, 47(9), 1092-1097 (2012-09-14)
In this work, we illustrate the application of proton transfer reaction mass spectrometry (PTR-MS) in the field of food and drink safety. We present proof-of-principle measurements of four different drinks (water, tea, red wine and white wine) each spiked separately
S R Graham et al.
The Medical journal of Australia, 149(11-12), 686-688 (1988-12-05)
The purpose of this review is to highlight the toxicity of chloral hydrate and to review the management of overdoses with chloral hydrate. Three patients are presented in whom life-threatening cardiac arrhythmias dominated the clinical presentation. These arrhythmias were resistant
Li Chen et al.
Developmental neurobiology, 74(9), 907-917 (2014-02-21)
Schizophrenia is a disease typically associated with an adolescent onset. Although there have been a considerable number of imaging studies investigating the transition to psychosis in prodromal patients, there are relatively few preclinical studies examining potential mechanisms that may contribute
Tal I Arnon et al.
PloS one, 3(5), e2150-e2150 (2008-05-15)
The natural cytotoxic receptors (NCRs) are a unique set of activating proteins expressed mainly on the surface of natural killer (NK) cells. The NCRs, which include three members; NKp46, NKp44 and NKp30, are critically involved in NK cytotoxicity against different
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service