XX1614700
Millipore MilliSolve Kit
47 mm, Bottle-to-Bottle Filtration System
Synonym(s):
Millipore Kit
About This Item
Recommended Products
material
PTFE tubing
anodized aluminum clamp(s)
borosilicate flask (with ground joint)
borosilicate glass base
borosilicate glass cap
borosilicate glass cover
borosilicate glass funnel(s)
silicone sleeve (for alignment)
stainless steel forceps
Quality Level
manufacturer/tradename
MilliSolve
technique(s)
vacuum filtration: suitable
diam.
12.5 cm
flask capacity
2 L
application(s)
filtration
shipped in
ambient
General description
47 mm, Bottle-to-Bottle Filtration System
The Millipore MilliSolve Filtration System has been designed for the filtration of liquid chromatography buffers and solvents under vacuum. The system uses a 0.45 µm membrane filter to eliminate particles which can shorten column life. Vacuum filtration with the Millipore MilliSolve system also removes a large portion of dissolved gasses from buffers, reducing the risk of air bubbles interfering with LC instruments. With automatic and continuous filtration, there is no need to add liquid into a funnel as filtration progresses. Filtration occurs in a closed system, which is important when filtering hazardous fluids. If open funnel filtration is required, the 300 mL funnel replaces the vacuum cover.
This item is a kit composed of the following items:
(Catalog Number - Description)
XX1614705 - Millipore Ground Joint Flask, 2L, Conical bottom
XX1514702 - Millipore Glass Base and Cap, 47 mm, Glass frit support
XX1614701 - Millipore Glass cover, 47 mm, For use with Millipore MilliSolve Kit
XX1014703 - Millipore Spring Clamp, 47 mm, Aluminum
XT1200000 - Millipore Tubing for Solvent/Buffer, PTFE, 70 cm
XX1014704 - Millipore Glass funnel, 300 mL, 47 mm, Borosilicate, Ground glass seal
ALIGNED05 - Millipore Alignment Guide, 47 mm, 5/PK
Features & Benefits:
• Automatic, continuous solvent and buffer filtration eliminates the need to pour during filtration
• 2L receiving flask includes plug and tubing for storage, eliminating filtered buffer transfer, reducing labor and lessening contamination risk
• Conical 2L flask bottom allows access to all filtered solvent or buffer
Applications:
Mobile Phase Preparation, Buffer Filtration
Principle
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service