Skip to Content
Merck
All Photos(2)

Key Documents

MABN2515

Sigma-Aldrich

Anti-Alpha Dystroglycan Antibody

clone 3D7

Synonym(s):

Anti Alpha-DGP, Anti-α-Dystroglycan Antibody, Anti-DAG, Dystroglycan Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

clone

monoclonal

species reactivity

mouse, human

technique(s)

western blot: suitable

General description

Dystroglycan (UniProt: Q62165; also known as Dystrophin-associated glycoprotein 1) is encoded by the Dag1 (also known as Dag-1) gene (Gene ID: 13138) in murine species. Dystroglycan is a component of the dystrophin glycoprotein complex that links the extracellular matrix to the intracellular actin cytoskeleton. It is synthesized with a signal peptide (aa 1-27) that is subsequently cleaved off to generate the mature protein that is further autolytically cleaved (between 651-652) to produce alpha-dystroglycan (aa 28-651) and beta-dystroglycan (aa 652-893). Alpha-dystroglycan is an extracellular peripheral glycoprotein that binds to several extracellular matrix and synaptic proteins such as laminin, agrin, neurexin, and pikachurin. It is reported that its O-mannosyl glycosylation is required for its ligand-binding functions. Beta-dystroglycan is a single-pass type 1 transmembrane protein that plays important roles in connecting the extracellular matrix to the cytoskeleton and serves as a cell adhesion receptor in both muscle and non-muscle tissues. In the cell, beta-dystroglycan binds to dystrophin that is linked to actin cytoskeleton. Hence, alpha/beta dystroglycans act as a molecular axis connecting extracellular matrix with the cytoskeleton across the plasma membrane. Defects in O-mannosyl glycan have been linked to various congenital muscular dystrophies caused by aberrant alpha-dystroglycan glycosylation. (Ref.: Sudo, A., et al. (2018). Human Mol. Genetics. 27(7); 7 1174-1185; Kanagawa, M., et al. (2010). J. Biol. Chem. 285(41); 31208-31216).

Immunogen

Murine alpha-dystroglycan-Fc-Fusion protein.

Application

Anti-Alpha Dystroglycan, clone 3D7, Cat. No. MABN2515, is a rat monoclonal antibody that detects Dystroglycan and has been tested for use in Western Blotting.
Western Blotting Analysis: A representative lot detected Alpha Dystroglycan in Western Blotting applications (Ohtsuka, Y., et. al. (2015). Sci Rep. 5:8316; Kanagawa, M., et. al. (2010). J Biol Chem. 285(41):31208-16; Kanagawa, M., et. al. (2016). Cell Rep

Quality

Evaluated by Western Blotting in mouse brain tissue lysates.

Western Blotting Analysis: 1 ug/mL of this antibody detected Alpha Dystroglycan in mouse brain tissue lysates.

Other Notes

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service