Skip to Content
Merck
All Photos(2)

Key Documents

MABN1796

Sigma-Aldrich

Anti-LRP1 Antibody, 85 kDa subunit Antibody, clone 6F8

clone 6F8, from mouse

Synonym(s):

Prolow-density lipoprotein receptor-related protein 1, A2MR, Alpha-2-macroglobulin receptor, APER, Apolipoprotein E receptor, CD19, Low-density lipoprotein receptor-related protein 1 85 kDa subunit, LRP-85

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6F8, monoclonal

species reactivity

human, mouse

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... LRP1(4035)

General description

Prolow-density lipoprotein receptor-related protein 1 (UniProt Q07954; also known as A2MR, Alpha-2-macroglobulin receptor, APER, Apolipoprotein E receptor, CD19. LRP-1) is encoded by the LRP1 (also known as A2MR, APR) gene (Gene ID 4035) in human. LRP-1 is single pass type I membrane protein that is expressed in most tissues, but is abundant in liver, brain, and lung. LRP1 is synthesized with a signal peptide sequence (a.a. 1-19) and is processed in trans-Golgi network by furin to generate a 515 kDa alpha subunit and an 85 kDa beta subunit. The alpha and beta subunits are non-covalently linked during LRP1 transport to the cell membrane. LRP1 recognizes and mediates the endocytosis of more than 40 different ligands, including apolipoprotein E (ApoE), APP and amyloid beta. It is the primary receptor mediating transport of amyloid beta peptides across the blood-brain barrier into circulation, thereby clearing them from the brain. LRP1 is required for early embryonic development and is involved in cellular lipid homeostasis and plasma clearance of chylomicron remnants and activated LRPAP1 (alpha 2-macroglobulin). Excessive copper accumulation in the brain has been linked with reduced LRP1 mediated clearance of amyloid beta peptides across the blood brain barrier, which may contribute to complications of Alzheimer′s disease.

Specificity

Clone 6F8 (A.k.a. 6AF8) epitope lies within LRP1 C-terminal end sequence present in the membrane-bound beta-fragment (85 kDa subunit), but not the extracellular alpha-fragment (515 kDa subunit). Target region is 100% conserved between human and murine species, but missing in human LRP1 spliced isoform 2.

Immunogen

Synthetic peptide corresponding to the C-terminal end sequence of human/mouse LRP1.

Application

Anti-LRP1, 85 kDa subunit, clone 6F8, Cat. No. MABN1796, is a highly specific mouse monoclonal antibody that targets LRP1 85 kDa subunit and has been tested in ELISA and Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected LRP1 85 kDa subunit in 10 µg of mouse hippocampus tissue lysate.

ELISA Analysis: A representative lot was employed as the capture antibody for the detection of LRP1 in different human brain regions by sandwich ELISA. a strong positive correlation between LRP1 and PSD95 regional distribution was observed (Shinohara, M., et al. (2013). Acta Neuropathol. 125(4):535-547).

Western Blotting Analysis: A representative lot detected siRNA-mediated LRP1 knockdown in human brain vascular pericytes (Casey, C.S., et al. 2015. J. Biol. Chem. 290(22):14208-14217).

Note: The use of 5% skim milk as the blocking agent and 1-2 hr instead of overnight primary incubation time is recommended for Western blotting application to minimize non-specific background.

Quality

Evaluated by Western Blotting in mouse brain and mouse hippocampus tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected LRP1 85 kDa subunit in 10 µg of mouse whole brain tissue lysate.

Target description

~85 kDa observed. Target band size appears larger than the calculated molecular weight of 65.78 kDa (a.a. 3944-4544; UniProt Q07954) due to glycosylation. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angela Jeong et al.
Acta neuropathologica communications, 9(1), 129-129 (2021-07-29)
The pathogenic mechanisms underlying the development of Alzheimer's disease (AD) remain elusive and to date there are no effective prevention or treatment for AD. Farnesyltransferase (FT) catalyzes a key posttranslational modification process called farnesylation, in which the isoprenoid farnesyl pyrophosphate
Dustin Chernick et al.
Journal of neurochemistry, 147(5), 647-662 (2018-07-22)
The apolipoprotein E (apoE) ε4 allele is the primary genetic risk factor for late-onset Alzheimer's disease (AD). ApoE in the brain is produced primarily by astrocytes; once secreted from these cells, apoE binds lipids and forms high-density lipoprotein (HDL)-like particles.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service