Skip to Content
Merck
All Photos(1)

Key Documents

MAB4625

Sigma-Aldrich

Anti-Bcl-X Antibody, clone 2H12

clone 2H12, Chemicon®, from mouse

Synonym(s):

Bcl-2-like Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2H12, monoclonal

species reactivity

rat, pig, mouse, human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BCL2L1(598)

General description

Bcl-X has two isoforms, Bcl-XL (long), a 241-aa protein, and Bcl-XS (short), a 178-aa protein lacking a 63 aa domain that is well conserved among members of the Bcl-2 family.

Specificity

MAB4625 recognizes a protein of 27 kDa, identified as the Bcl-X protein. Reacts with both Bcl-XS and Bcl-XL.

Immunogen

Synthetic peptide, aa 3-14 of human Bcl-X protein. This amino acid sequence is shared by human and murine Bcl-X protein.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
BCL2 & Inhibition
This Anti-Bcl-X Antibody, clone 2H12 is validated for use in FC, IF, IP, WB, IH for the detection of Bcl-X.
Wstern Blot: 1μg/mL (Hsu, 1997a; Hsu, 1997b)

Flow Cytometry

Immunofluorescence Immunoprecipitation: native protein (2ug/mg protein lysate).

Immunohistochemistry (frozen and paraffin): 2-4ug/mL for 30 minutes at room temperature.

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Protein A purified immunoglobulin in 10mM PBS, pH 7.4, with 0.2% BSA and 0.09% sodium azide.

Storage and Stability

Maintain at 2-8°C for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilka Knippertz et al.
Journal of immunology research, 2016, 6078473-6078473 (2016-07-23)
To specifically target dendritic cells (DCs) to simultaneously express different therapeutic transgenes for inducing immune responses against tumors, we used a combined promoter system of adenoviral vectors. We selected a 216 bp short Hsp70B' core promoter induced by a mutated, constitutively
Vassiliki Poulaki et al.
Investigative ophthalmology & visual science, 48(10), 4706-4719 (2007-09-28)
To evaluate the potential of proteasome inhibitors, a novel class of antitumor agents, for the treatment of retinoblastoma. The proteasome inhibitor bortezomib (PS-341, Velcade; Millennium Pharmaceuticals, Cambridge, MA), approved by the US Food and Drug Administration for the treatment of
Carles Solà-Riera et al.
PLoS pathogens, 16(2), e1008297-e1008297 (2020-02-08)
Hantaviruses, zoonotic RNA viruses belonging to the order Bunyavirales, cause two severe acute diseases in humans, hemorrhagic fever with renal syndrome (HFRS) and hantavirus pulmonary syndrome (HPS). Hantavirus-infected patients show strong cytotoxic lymphocyte responses and hyperinflammation; however, infected cells remain
Yizhi Zhang et al.
International journal of molecular medicine, 40(4), 1089-1095 (2017-08-03)
Grifola frondosa, a type of food and medical fungus, has been shown to exhibit various pharmacological activities, including anticancer effects. As the most typical cancer diagnosed among female patients, breast cancer remains a huge concern threatening human health globally. In the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service