Skip to Content
Merck
All Photos(1)

Key Documents

MAB371

Sigma-Aldrich

Anti-Cranin Antibody, aa572-604 of human sequence, clone 6C1

ascites fluid, clone 6C1, Chemicon®

Synonym(s):

alpha-dystroglycan

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
6C1, monoclonal
application:
ELISA
ICC
IHC
species reactivity:
mouse, sheep, rat, human, chicken, Xenopus
technique(s):
ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
citations:
1

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

6C1, monoclonal

species reactivity

mouse, sheep, rat, human, chicken, Xenopus

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgM

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... DAG1(1605)

Specificity

Cranin (dystroglycan), extracellular site, near the C-terminus of the alpha-subunit. The expression and localization of cranin varies considerably according to the cells physiologic state and differentiaion state. MAB371 recognizes the same peptide region as MAB373 but a different epitope.

Immunogen

Epitope: aa572-604 of human sequence
Synthetic peptide corresponding to a.a. 572-604 of human cranin.

Application

Immunohistochemistry on tissue fixed briefly with 0.5% glutaraldehyde, 2% PLP fixation, methanol or acetone. Prolonged fixation must be avoided. Traditional formalin is not recommended. Suggested working dilution 1:1-1:50.

Immunocytochemistry on neuron, astrocyte, smooth muscle, fibroblast and epithelial cell lines.

ELISA

Not suggested for use in Western blot.

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure

Cell Structure
Research Sub Category
Cytoskeleton

ECM Proteins
This Anti-Cranin Antibody, aa572-604 of human sequence, clone 6C1 is validated for use in ELISA, IC, IH for the detection of Cranin.

Physical form

Ascites Liquid.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months from date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Li et al.
The Journal of cell biology, 153(4), 811-822 (2001-05-16)
Fibroblast growth factors and receptors are intimately connected to the extracellular matrix by their affinity to heparan sulfate proteoglycans. They mediate multiple processes during embryonic development and adult life. In this study, embryonic stem cell-derived embryoid bodies were used to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service