Skip to Content
Merck
All Photos(1)

Key Documents

DX2370

Supelco

Dithizone

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Dithizone, Diphenylthiocarbazone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHNHCSN=NC6H5
CAS Number:
MDL number:
UNSPSC Code:
12352127
EC Index Number:
200-454-1
Pricing and availability is not currently available.

Agency

meets USP/NF

Assay

≥85.0% (spectrophotometric assay)

form

powder or crystals

impurities

≤0.002% Heavy metals (as Pb)

ign. residue

≤0.2%

color

black

mp

168 °C (decomposition)

shipped in

ambient

storage temp.

room temp

SMILES string

S=C(NNc2ccccc2)N=Nc1ccccc1

InChI

1S/C13H12N4S/c18-13(16-14-11-7-3-1-4-8-11)17-15-12-9-5-2-6-10-12/h1-10,14H,(H,16,18)

InChI key

UOFGSWVZMUXXIY-UHFFFAOYSA-N

Application

Dithizone is a high purity product that meets reagent specifications suitable for testing USP/NF monographs.

Analysis Note

Assay (spectrophotometric): 85.0% min
Color: Black
Form: Fine crystals or powder
Heavy metals (as Pb): 0.002% max
Identity: Passes test
Ratio of absorbances: 1.55 min
Residue after ignition: 0.2% max

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service