Skip to Content
Merck
All Photos(5)

Documents

BPCKHF09004S25

Millipore

Hi-Flow Plus Membrane 90

4mil backing, 25mm width, pack of 11 slit rolls

Synonym(s):

HF, Hi-Flow, immunoassay membrane, lateral flow membrane, nitrocellulose membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NA.55

Quality Level

packaging

pack of 11 roll

manufacturer/tradename

Hi-Flow Plus

parameter

90 sec/4 cm capillary flow rate

backing thickness

4 mil (100 μm)

filter W × L

25 mm × 100 m

roll L

100 m (328 ft)

slit W

25 mm

Looking for similar products? Visit Product Comparison Guide

General description

Hi-Flow Plus membranes are designed and optimized for use in rapid diagnostic lateral flow tests that require superior consistency and performance. Lateral flow tests facilitate quick and convenient detection/quantification of analytes such as antibodies and antigens present in biological samples, including blood, saliva, and urine. Hi-Flow Plus membranes are ideally suited for such applications. Available in a variety of capillary flow times and backing options, choosing Hi-Flow Plus membranes can improve assay performance and offer flexibility to meet a wide range of assay design requirements.

Hi-Flow Plus 90 (HF090) is suitable for tests where speed is critical, the sample is plentiful, and sensitivity is not a concern. This Hi-Flow Plus 90 pack comprises 11 slit rolls of 4 mil backed nitrocellulose membrane with a nominal capillary flow time of 90 seconds/4 cm.

Application

  • Hi-Flow Plus membrane is a critical raw material suitable for use in a broad range of Lateral Flow test applications, including Infectious Disease testing (COVID-19, HIV, Influenza, Malaria, STDs, etc.), as well as in women’s health, biomarker detection, drugs of abuse testing, drug development, environmental testing, agricultural applications, food safety, product quality testing, and animal health.
  • It is suitable for use with all lateral flow test techniques including competitive, sandwich, and multiplex detection assays.
  • It is optimized for use in both qualitative and quantitative test types and pairs well with lateral flow detector particles including colloidal gold nanoparticles, dyed/fluorescent microspheres, etc.
  • It is suitable for use in lateral flow tests designed to detect analytes present in a broad range of sample matrices including, blood, saliva, urine, nasopharyngeal swabs, sweat, and others.

Features and Benefits

FEATURES:
  • Consistent flow and proprietary surface chemistry optimized for lateral flow applications.
  • Robust manufacturing process.
  • High Quality Level: MQ400.
  • Trusted Partner and brand with decades of membrane manufacturing experience.
  • High production capacity, high stock levels, multiple manufacturing lines.
  • Convenient sample packs available in several formats.
  • Manufacturing scale quantities available to purchase online with immediate shipping.
  • We offer a complete solution; along with Hi-Flow Plus membranes we offer raw materials across the entire lateral flow workflow as well as assay development services: https://www.sigmaaldrich.com/US/en/services/ivd-development-manufacturing/lateral-flow-services.
BENEFITS:
  • Consistent capillary flow time and membrane thickness.
  • Superior surface morphology allows for consistent and even flow distribution of reagents across the membrane surface.
  • Sharp, intense, and uniform test and control line formation.
  • Excellent intra-lot and inter-lot consistency.
  • Better assay sensitivity.
  • Excellent reproducibility.
  • Improved lateral flow test manufacturing yields.
  • Quick delivery and lead time.
  • No supply disruption.
  • Excellent technical support.

Legal Information

Hi-Flow is a trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lateral flow assays
Katarzyna M Koczula, Andrea Gallotta
Essays in Biochemistry, 60(1), 111-120 (2016)

Articles

Explore design, materials, & manufacturing of lateral flow tests. Learn about reproducibility, optimization, & Hi-Flow™ Plus membranes for efficient scale-up.

Explore design, materials, & manufacturing of lateral flow tests. Learn about reproducibility, optimization, & Hi-Flow™ Plus membranes for efficient scale-up.

Explore design, materials, & manufacturing of lateral flow tests. Learn about reproducibility, optimization, & Hi-Flow™ Plus membranes for efficient scale-up.

Explore design, materials, & manufacturing of lateral flow tests. Learn about reproducibility, optimization, & Hi-Flow™ Plus membranes for efficient scale-up.

Related Content

Discover critical membrane materials for your IVD lateral flow immunoassay development including Hi-Flow™ lateral flow membrane cards and SureWick® pad materials.

Discover critical membrane materials for your IVD lateral flow immunoassay development including Hi-Flow™ lateral flow membrane cards and SureWick® pad materials.

Discover critical membrane materials for your IVD lateral flow immunoassay development including Hi-Flow™ lateral flow membrane cards and SureWick® pad materials.

Discover critical membrane materials for your IVD lateral flow immunoassay development including Hi-Flow™ lateral flow membrane cards and SureWick® pad materials.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service