Skip to Content
Merck
All Photos(1)

Key Documents

8.52120

Sigma-Aldrich

Fmoc-Gln(Dmcp)-OH

Novabiochem®

Synonym(s):

Fmoc-Gln(Dmcp)-OH, N-α-Fmoc-N-γ-(1-cyclopropyl-1-methylethyl)-L-glutamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H30N2O5
CAS Number:
Molecular Weight:
450.53
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

Quality Level

product line

Novabiochem®

Assay

≥90.0% (acidimetric)
≥98% (TLC)
≥98.0% (HPLC)

form

solid

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amide

storage temp.

15-25°C

SMILES string

N([C@@H](CCC(=O)NC(C4CC4)(C)C)C(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C26H30N2O5/c1-26(2,16-11-12-16)28-23(29)14-13-22(24(30)31)27-25(32)33-15-21-19-9-5-3-7-17(19)18-8-4-6-10-20(18)21/h3-10,16,21-22H,11-15H2,1-2H3,(H,27,32)(H,28,29)(H,30,31)/t22-/m0/s1

InChI key

JHXHPPOYVOMYCD-QFIPXVFZSA-N

General description

A superior derivative to Fmoc-Gln(Trt)-OH for the synthesis of Gln-containing peptides by Fmoc SPPS. Fmoc-Gln(Dmcp)-OH is more soluble in DMF than Fmoc-Gln(Trt)-OH, thereby facilitating coupling reactions at higher concentration. Cleavage of the Dmcp group is rapid, even when the residue is located at the N-terminus of a peptide. Coupling of Dmcp-protected derivatives is faster than that of the corresponding hindered Trt derivatives. Dmcp-protected peptides have enhanced solubility.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Literature references

[1] L. A. Carpino, et al. (1995) J. Org. Chem., 60, 7718.

Linkage

Replaces: 04-12-1292

Analysis Note

Assay (acidimetric): ≥ 90.0 %
Assay (HPLC, area%): ≥ 98.0 % (a/a)
Water (K. F.): ≤ 1.00 %
Identity (IR): passes test
Solubility (1 mmole in 2 ml DMF): passes test
Color (visual): passes test
Appearance of substance (visual): passes test
Purity (TLC(CMA2)): ≥ 98 %
Enantiomeric purity: ≥ 99.5 % (a/a)

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service