Skip to Content
Merck
All Photos(2)

Key Documents

8.51009

Sigma-Aldrich

PyBOP®

greener alternative

Novabiochem®

Synonym(s):

Benzotriazole-1-yl-oxy-tris-pyrrolidino-phosphonium hexafluorophosphate, PyBOP®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H28N6OP · F6P
CAS Number:
Molecular Weight:
520.39
MDL number:
UNSPSC Code:
12352107
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

product line

Novabiochem®

Assay

≥99.0% (HPLC)

form

powder

reaction suitability

reaction type: Coupling Reactions

manufacturer/tradename

Novabiochem®

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Learn more about the Principles of Green Chemistry.

mp

140-155 °C

application(s)

peptide synthesis

greener alternative category

storage temp.

2-8°C

SMILES string

[P+](N5CCCC5)(N4CCCC4)(N3CCCC3)O[n]1nnc2c1cccc2

InChI

1S/C18H28N6OP/c1-2-10-18-17(9-1)19-20-24(18)25-26(21-11-3-4-12-21,22-13-5-6-14-22)23-15-7-8-16-23/h1-2,9-10H,3-8,11-16H2/q+1

InChI key

WGNZRLMOMHJUSP-UHFFFAOYSA-N

General description

The standard in situ coupling reagent for solid phase peptide synthesis [1,2,3,4,5]. This product can replace BOP in all applications without loss of performance. Unlike uronium-based reagents, PyBOP® does not give rise to guanidinated by-products during cyclization and fragment condensation reactions.

Associated Protocols and Technical Articles
Guide to Selection of Coupling Reagents

Literature references

[1] J. Martinez, et al. (1985) J. Med. Chem., 28, 1874.
[2] J. Coste, et al. (1990) Tetrahedron Lett., 31, 205.
[3] G. B. Fields, et al. in ′Innovation & Perspectives in Solid Phase Synthesis, 1st International Symposium′, R. Epton (Eds), SPCC UK Ltd., Birmingham, 1990, pp. 241.
[4] T. Høeg-Jensen, et al. (1991) Tetrahedron Lett., 32, 7617.
[5] R. von Eggelkraut-Gottanka, et al. (2003) Tetrahedron Lett., 44, 3551.
We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. PyBOP is widely used as a substitute for the BOP, with the aim of avoiding formation of hexamethylphosphoric triamide (HMPA) which is a carcinogenic side product. Click here for more information.

Linkage

Replaces: 01-62-0016

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Assay (HPLC, area%): ≥ 99.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany
PyBOP is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Novabiochem® coupling reagents for in situ activation are fast-reacting and compatible with various amino acids.

Novabiochem® coupling reagents for in situ activation are fast-reacting and compatible with various amino acids.

Novabiochem® coupling reagents for in situ activation are fast-reacting and compatible with various amino acids.

Novabiochem® coupling reagents for in situ activation are fast-reacting and compatible with various amino acids.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service