Skip to Content
Merck
All Photos(1)

Key Documents

8.21903

Sigma-Aldrich

4-Methylbenzoic acid

for synthesis

Synonym(s):

4-Methylbenzoic acid, p-Toluic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(CH3)C6H4COOH
CAS Number:
Molecular Weight:
136.15
MDL number:
UNSPSC Code:
12352106
EC Index Number:
202-803-3

vapor pressure

0.02 hPa ( 70 °C)

Quality Level

Assay

≥98.0% (HPLC)

form

powder

autoignition temp.

570 °C

potency

400 mg/kg LD50, oral (Rat)

bp

274 °C/1013 hPa

mp

179-181 °C

transition temp

flash point 181 °C

solubility

0.3 g/L

density

1.06 g/cm3 at 20 °C

bulk density

700 kg/m3

storage temp.

2-30°C

InChI

1S/C8H8O2/c1-6-2-4-7(5-3-6)8(9)10/h2-5H,1H3,(H,9,10)

InChI key

LPNBBFKOUUSUDB-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 179 °C
Melting range (upper value): ≤ 182 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

357.8 °F

Flash Point(C)

181 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service