Skip to Content
Merck
All Photos(2)

Key Documents

8.20423

Sigma-Aldrich

N,N′-Dibutylthiourea

for synthesis

Synonym(s):

N,N′-Dibutylthiourea

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2NH)2CS
CAS Number:
Molecular Weight:
188.33
MDL number:
UNSPSC Code:
12352111
EC Index Number:
203-674-6
NACRES:
NA.22
form:
powder

vapor pressure

0.0032 hPa ( 25 °C)

Quality Level

form

powder

potency

350 mg/kg LD50, oral (Rat)

pH

6 (20 °C, 0.2 g/L in H2O)

mp

62-65 °C

bulk density

500 kg/m3

storage temp.

2-30°C

SMILES string

S=C(NCCCC)NCCCC

InChI

1S/C9H20N2S/c1-3-5-7-10-9(12)11-8-6-4-2/h3-8H2,1-2H3,(H2,10,11,12)

InChI key

KFFQABQEJATQAT-UHFFFAOYSA-N

Analysis Note

Assay (of S): ≥ 98.0 %
Melting range (lower value): ≥ 63 °C
Melting range (upper value): ≤ 66 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service