Skip to Content
Merck
All Photos(2)

Key Documents

8.20161

Sigma-Aldrich

4,4′-Bis(diethylamino)-benzophenone

for synthesis

Synonym(s):

4,4′-Bis(diethylamino)-benzophenone, N,N,N′,N′-Tetraethyl-4,4′-diaminobenzophenone, Michler ethylketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-[C2H5)2N]C6H4COC6H4-4-[N(C2H5)2]
CAS Number:
Molecular Weight:
324.46
MDL number:
UNSPSC Code:
12352115
EC Index Number:
202-025-4
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (HPLC)

form

powder

potency

5000 mg/kg LD50, oral (Rat)

pH

4-6 (20 °C, 50 g/L in H2O)

mp

90-95 °C

transition temp

flash point 151 °C

bulk density

400 kg/m3

storage temp.

2-30°C

SMILES string

N(CC)(CC)c1ccc(cc1)C(=O)c2ccc(cc2)N(CC)CC

InChI

1S/C21H28N2O/c1-5-22(6-2)19-13-9-17(10-14-19)21(24)18-11-15-20(16-12-18)23(7-3)8-4/h9-16H,5-8H2,1-4H3

InChI key

VYHBFRJRBHMIQZ-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 92 °C
Melting range (upper value): ≤ 96 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service