Skip to Content
Merck
All Photos(2)

Key Documents

8.07103

Sigma-Aldrich

Palladium(II) acetylacetonate

≥98% (ICP)

Synonym(s):

Palladium(II) acetylacetonate, Bis(acetylacetonato)palladium(II), Bis(2,4-pentanedionato) palladium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O4Pd
CAS Number:
Molecular Weight:
304.64
MDL number:
UNSPSC Code:
12352103
EC Index Number:
237-859-8

Assay

≥98% (ICP)

form

powder

storage temp.

2-30°C

General description

for synthesis

Analysis Note

Assay (ICP-OES): ≥ 98.0 %

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service