Skip to Content
Merck
All Photos(1)

Key Documents

8.07024

Sigma-Aldrich

Propiophenone

for synthesis

Synonym(s):

Propiophenone, 1-Phenyl-1-propanone, Ethyl phenyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O
CAS Number:
Molecular Weight:
134.18
MDL number:
UNSPSC Code:
12352115
EC Index Number:
202-257-6
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

1.99 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

potency

4490 mg/kg LD50, oral (Rat)
4490 mg/kg LD50, skin (Rabbit)

bp

214-218 °C/1013 hPa

mp

18 °C

transition temp

flash point 85 °C

solubility

2 g/L

density

1.01 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O=C(CC)c1ccccc1

InChI

1S/C9H10O/c1-2-9(10)8-6-4-3-5-7-8/h3-7H,2H2,1H3

InChI key

KRIOVPPHQSLHCZ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.009 - 1.010
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service