8.02189
tert-Butyl acetate
for synthesis
Synonym(s):
tert-Butyl acetate, Acetic acid tert-butyl ester
About This Item
Recommended Products
vapor pressure
41 hPa ( 20 °C)
Quality Level
Assay
≥99.0% (GC)
form
liquid
potency
4100 mg/kg LD50, oral (Rat)
expl. lim.
1.5 % (v/v)
bp
95-97 °C/1013 hPa
transition temp
flash point 1 °C
density
0.87 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
O(C(C)(C)C)C(=O)C
InChI
1S/C6H12O2/c1-5(7)8-6(2,3)4/h1-4H3
InChI key
WMOVHXAZOJBABW-UHFFFAOYSA-N
Analysis Note
Density (d 20 °C/ 4 °C): 0.865 - 0.866
Identity (IR): passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Flam. Liq. 2
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
39.2 °F - closed cup
Flash Point(C)
4 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service