Skip to Content
Merck
All Photos(1)

Key Documents

4.85085

Supelco

Formic Acid Solution

0.1% (v/v) Formic Acid Mobile Phase in Water

Synonym(s):

FA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
50202301
bp:
100 °C/1013 hPa

Product Name

Water/Formic Acid 0.1% Mobile Phase,

form

liquid

Quality Level

shelf life

12 mo. (from manufacture date)

contains

0.095-0.105 % (v/v) trifluoroacetic acid

bp

100 °C/1013 hPa

density

~0.9985 g/cm3 at 20 °C

UV absorption

λ: 220 nm Amax: ≤1.0
λ: 230 nm Amax: ≤0.5
λ: 240 nm Amax: ≤0.10
λ: 250 nm Amax: ≤0.02
λ: 280 nm Amax: ≤0.01

shipped in

ambient

Application

4.85085.4000 is packaged in poly coated glass bottles

Analysis Note

Expiration date12 months for date of maufactureAssay (HCOOH)0.095-0.105% v/vDensity at 20°Capproximately 0.9985g/cm³FormClear liquidGradient at 254 nm0.005AU maxGradient at 280 nm0.003AU maxUV Absorbance at 220 nm1.0AU maxUV Absorbance at 230 nm0.5AU maxUV Absorbance at 240 nm0.10AU maxUV Absorbance at 250 nm0.02AU maxUV Absorbance at 280 nm0.01AU max

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service