407710
α-Iodoacetamide
≥99% (HPLC), crystalline solid, cysteine protease inhibitor, Calbiochem®
Synonym(s):
α-Iodoacetamide
About This Item
Recommended Products
Product Name
α-Iodoacetamide, α-Iodoacetamide, CAS 144-48-9, is an irreversible inhibitor of several cysteine proteases.
Quality Level
Assay
≥99% (HPLC)
form
crystalline solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
white
solubility
methanol: 1:20
DMF: soluble
ethanol: soluble
water: soluble
shipped in
ambient
storage temp.
−20°C
SMILES string
ICC(=O)N
InChI
1S/C2H4INO/c3-1-2(4)5/h1H2,(H2,4,5)
InChI key
PGLTVOMIXTUURA-UHFFFAOYSA-N
General description
Biochem/physiol Actions
cysteine proteases
Warning
Reconstitution
Other Notes
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Aquatic Chronic 4 - Resp. Sens. 1 - Skin Sens. 1
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service